Assay Details
Target Gene Details
Entrez Gene ID: | 17183 |
Gene Name: | matrilin 4 |
Gene Aliases: |
matrilin-4 |
Location: |
Chr.2:164389393-164405160 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Matn4 | NM_001252563.1 | 8 | 1391 | NP_001239492.1 |
NM_013592.4 | 8 | 1656 | NP_038620.2 | |
AJ006140.1 | 8 | 1375 | CAA06889.1 | |
AK041073.1 | 8 | 1364 | ||
BC036558.1 | 8 | 1391 | AAH36558.1 |
More Information
Set Membership: |
Intragenic Exonic |