Assay Details
Target Gene Details
Entrez Gene ID: | 18019 |
Gene Name: | nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2 |
Gene Aliases: |
AI607462, NF-ATc2, NF-ATp, NFAT1, NFAT1-D, Nfatp |
Location: |
Chr.2:168476410-168601657 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
More Information
Set Membership: |
Intragenic Exonic |