Assay Details
Target Gene Details
Entrez Gene ID: | 11538 |
Gene Name: | activity-dependent neuroprotective protein |
Gene Aliases: |
AA589558, mKIAA0784 |
Location: |
Chr.2:168180965-168207062 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adnp | NM_009628.2 | 1 | 87 | NP_033758.2 |
AK129214.1 | 1 | 137 | BAC98024.1 |