Assay Details
Target Gene Details
Entrez Gene ID: | 19099 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 1 |
Gene Aliases: |
IB1, JIP-1, Jip1, Prkm8ip, Skip, mjip-2a |
Location: |
Chr.2:92383676-92401263 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mapk8ip1 | NM_001202445.1 | 12 | 2735 | NP_001189374.1 |
NM_001202446.1 | 12 | 2660 | NP_001189375.1 | |
NM_011162.5 | 12 | 2443 | NP_035292.2 | |
AF003115.1 | 12 | 2302 | AAB66317.1 | |
AF054611.1 | 12 | 2443 | AAD22580.1 | |
AF109768.1 | AAD38346.1 | |||
AF109769.1 | AAD38347.1 | |||
AF109770.1 | AAD38348.1 | |||
AF109771.1 | AAD38349.1 | |||
AK147033.1 | 12 | 2662 | BAE27625.1 | |
BC072578.1 | 12 | 2627 | AAH72578.1 |
More Information
Set Membership: |
Intragenic Exonic |