Assay Details
Target Gene Details
Entrez Gene ID: | 17005 |
Gene Name: | leukocyte tyrosine kinase |
Gene Aliases: |
- |
Location: |
Chr.2:119751326-119760431 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ltk | NM_008523.2 | 14 | 2028 | NP_032549.2 |
NM_203345.2 | 20 | 2874 | NP_976220.2 | |
NM_206941.1 | 15 | 2211 | NP_996824.1 | |
NM_206942.2 | 19 | 2691 | NP_996825.2 | |
AK157478.1 | 12 | 2025 | ||
BC047138.1 | 14 | 2028 | ||
BC141299.1 | 19 | 2667 | ||
M90470.1 | 20 | 2874 | AAA39451.1 | |
X07984.1 | 14 | 2002 | CAA30793.1 | |
X52621.1 | 14 | 2017 | CAA36848.1 |
More Information
Set Membership: |
Intragenic Exonic |