Assay Details
Target Gene Details
Entrez Gene ID: | 69601 |
Gene Name: | disabled 2 interacting protein |
Gene Aliases: |
2310011D08Rik, AI480459, Aip1, mKIAA1743 |
Location: |
Chr.2:35558458-35730994 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dab2ip | NM_001001602.2 | 4 | 948 | NP_001001602.2 |
NM_001114124.2 | 6 | 1072 | NP_001107596.1 | |
NM_001114125.1 | 6 | 924 | NP_001107597.1 | |
NM_001290637.1 | 6 | 979 | NP_001277566.1 | |
NM_001290639.1 | 4 | 607 | NP_001277568.1 | |
NM_001290640.1 | 4 | 912 | NP_001277569.1 | |
NM_001290641.1 | 6 | 1205 | NP_001277570.1 | |
AK147464.1 | 6 | 1004 | BAE27930.1 | |
AK147593.1 | 6 | 1206 | BAE28013.1 | |
AK155296.1 | 6 | 980 | BAE33173.1 | |
AY305656.1 | 4 | 665 | AAQ77379.1 | |
AY305657.1 | 4 | 948 | AAQ77380.1 | |
AY305658.1 | 4 | 607 | AAQ77381.1 | |
BC118530.1 | 4 | 573 | ||
DQ473307.1 | 6 | 809 |
More Information
Set Membership: |
Intragenic Exonic |