Assay Details
Target Gene Details
Entrez Gene ID: | 210356 |
Gene Name: | NCK-associated protein 5 |
Gene Aliases: |
8430408F21, D130011D22Rik, E030049G20Rik, Erih1, Erih2, Gm1548, Nap5 |
Location: |
Chr.1:125913636-126830897 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nckap5 | NM_001081756.1 | 15 | 5586 | NP_001075225.1 |
NM_172484.3 | 13 | 5447 | NP_766072.2 | |
NM_176957.3 | 12 | 1476 | NP_795931.2 | |
AK053243.1 | 10 | 1159 | BAC35319.1 | |
AY957601.1 | 10 | 4814 | AAY23348.1 | |
AY957602.1 | 11 | 5006 | AAY23349.1 | |
BC066095.1 | 13 | 5447 | AAH66095.1 | |
BC079551.1 | 13 | 5447 | AAH79551.1 |