Assay Details
Target Gene Details
Entrez Gene ID: | 15064 |
Gene Name: | major histocompatibility complex, class I-related |
Gene Aliases: |
H2ls |
Location: |
Chr.1:155127872-155146813 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mr1 | NM_008209.4 | 6 | 1558 | NP_032235.1 |
AF010448.1 | 6 | 1469 | AAD01444.1 | |
AF010449.1 | 6 | 1284 | ||
AF010450.1 | 6 | 1284 | ||
AF010451.1 | 3 | 2253 | ||
AK042119.1 | BAC31174.1 | |||
AK137171.1 | 6 | 1399 | ||
AK144040.1 | ||||
BC026137.1 | 6 | 1586 | AAH26137.1 |
More Information
Set Membership: |
Intragenic Exonic |