Assay Details
Target Gene Details
Entrez Gene ID: | 241070 |
Gene Name: | G protein-coupled receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.1:63182571-63223278 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpr1 | NM_146250.2 | 3 | 513 | NP_666362.1 |
BC032934.1 | 3 | 441 | AAH32934.1 |
More Information
Set Membership: |
Intragenic Exonic |