Assay Details
Target Gene Details
Entrez Gene ID: | 19264 |
Gene Name: | protein tyrosine phosphatase, receptor type, C |
Gene Aliases: |
B220, CD45R, Cd45, L-CA, Ly-5, Lyt-4, T200, loc |
Location: |
Chr.1:138062859-138175755 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprc | NM_001111316.2 | 32 | 5251 | NP_001104786.2 |
NM_001268286.1 | 29 | 4860 | NP_001255215.1 | |
NM_011210.4 | 30 | 4932 | NP_035340.3 | |
AK079399.1 | 1 | 1035 | ||
BM239381.2 | 1 | 318 |
More Information
Set Membership: |
Intragenic Exonic |