Assay Details
Target Gene Details
Entrez Gene ID: | 98752 |
Gene Name: | Fc receptor-like A |
Gene Aliases: |
BB219290, FCRL, FCRL1, FREB, Fcrlm1, Fcrx, Freb1, mFREB, mFcrX |
Location: |
Chr.1:170917594-170927590 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcrla | NM_001160215.1 | 4 | 769 | NP_001153687.1 |
NM_145141.2 | 4 | 766 | NP_660123.1 | |
NR_027666.1 | 4 | 835 | ||
AF329487.1 | 4 | 733 | AAL23897.1 | |
AF426462.1 | 4 | 726 | AAL58112.1 | |
AF531424.1 | 4 | 755 | AAM97592.1 | |
AK041572.1 | 4 | 746 | BAC30990.1 | |
AK172370.1 | 4 | 759 | BAE42968.1 | |
BC064708.1 | 4 | 769 | AAH64708.1 |
More Information
Set Membership: |
Intragenic Exonic |