Assay Details
Target Gene Details
Entrez Gene ID: | 14125 |
Gene Name: | Fc receptor, IgE, high affinity I, alpha polypeptide |
Gene Aliases: |
FcERI, Fce1a, Fcr-5, fcepsilonri |
Location: |
Chr.1:173221269-173227232 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcer1a | NM_010184.2 | 5 | 903 | NP_034314.2 |
BC125455.1 | ||||
BC125457.1 | ||||
BY585428.1 | 2 | 313 | ||
J05018.1 | 5 | 888 | AAA37600.1 |
More Information
Set Membership: |
Intragenic Exonic |