Assay Details
Target Gene Details
Entrez Gene ID: | 66385 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 7 |
Gene Aliases: |
2310014J01Rik, SDS22 |
Location: |
Chr.1:93343645-93367619 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r7 | NM_023200.2 | 10 | 2287 | NP_075689.1 |
AF222867.1 | 10 | 2287 | AAK00624.1 | |
AK129024.1 | 10 | 2304 | ||
AK145136.1 | 10 | 2293 | BAE26253.1 | |
AK162517.1 | 10 | 2295 | BAE36954.1 |
More Information
Set Membership: |
Intragenic Exonic |