Assay Details
Target Gene Details
Entrez Gene ID: | 67444 |
Gene Name: | integrin-linked kinase-associated serine/threonine phosphatase 2C |
Gene Aliases: |
0710007A14Rik, 1600009O09Rik, PP2C-DELTA |
Location: |
Chr.1:91373846-91398841 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ilkap | NM_023343.2 | 12 | 1287 | NP_075832.1 |
AK003007.1 | 11 | 2260 | ||
AK019158.1 | 6 | 671 | BAB31574.1 | |
BC026953.1 | 12 | 1287 | AAH26953.1 | |
BC027439.1 |
More Information
Set Membership: |
Intragenic Exonic |