Assay Details
Target Gene Details
Entrez Gene ID: | 13349 |
Gene Name: | atypical chemokine receptor 1 (Duffy blood group) |
Gene Aliases: |
AA162249, CCBP1, CD234, Darc, Dfy, ESTM35, FY, GPD |
Location: |
Chr.1:173331886-173333503 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ackr1 | NM_010045.2 | 2 | 982 | NP_034175.2 |
AF016584.1 | 2 | 923 | AAC53354.1 | |
AF109159.1 | 2 | 982 | AAF14225.1 | |
AY289754.1 | 2 | 889 | AAP42463.1 | |
AY291295.1 | 2 | 889 | AAP74548.1 | |
BC005583.1 | 2 | 981 | AAH05583.1 |
More Information
Set Membership: |
Intragenic Exonic |