Assay Details
Target Gene Details
Entrez Gene ID: | 111175 |
Gene Name: | peroxisomal trans-2-enoyl-CoA reductase |
Gene Aliases: |
2400003B18Rik, TERP |
Location: |
Chr.1:72259172-72284314 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pecr | NM_023523.5 | 8 | 965 | NP_076012.3 |
AF232011.1 | 8 | 871 | AAF69800.1 | |
AF242204.1 | 8 | 905 | AAK28336.1 | |
AK010260.1 | 8 | 956 | BAB26803.1 | |
AK090327.1 | 8 | 894 | ||
AK162100.1 | 8 | 914 | ||
BC013530.1 | 8 | 889 | AAH13530.1 | |
CB723327.1 | 3 | 181 |
More Information
Set Membership: |
Intragenic Exonic |