Assay Details
Target Gene Details
Entrez Gene ID: | 100169864 |
Gene Name: | predicted gene, 38419 |
Gene Aliases: |
Slc10a3-ubl4, Tu11 |
Location: |
Chr.X:74367447-74373349 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm38419 | NM_001278271.1 | 4 | 675 | NP_001265200.1 |
NM_001278272.1 | 4 | 639 | NP_001265201.1 | |
NR_103489.1 | 4 | 675 | ||
EU599043.1 | 4 | 544 | ||
EU599044.1 | 4 | 508 | ||
EU599045.1 | 4 | 544 |
Target Gene Details
Entrez Gene ID: | 27643 |
Gene Name: | ubiquitin-like 4 |
Gene Aliases: |
DXHXS254E, DXS254E, DXS254Eh, Gdx, Ubl4a |
Location: |
Chr.X:74365718-74368548 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubl4 | NM_145405.2 | 1 | 76 | NP_663380.1 |
NR_045779.1 | 1 | 76 | ||
AJ627038.1 | CAF25307.1 | |||
AK048689.1 | ||||
BC010817.1 | 1 | 55 | AAH10817.1 | |
BM932729.1 | 1 | 50 | ||
BY107969.1 | 1 | 47 | ||
BY273712.1 | 1 | 77 |