Assay Details
Target Gene Details
Entrez Gene ID: | 22201 |
Gene Name: | ubiquitin-like modifier activating enzyme 1 |
Gene Aliases: |
A1S9, Sbx, Ube-1, Ube1x |
Location: |
Chr.X:20658302-20683179 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uba1 | NM_001136085.2 | NP_001129557.1 | ||
NM_001276316.1 | NP_001263245.1 | |||
NM_001276317.1 | NP_001263246.1 | |||
NM_009457.4 | NP_033483.2 | |||
AK081808.1 | ||||
AK088057.1 | BAC40121.1 | |||
AK088528.1 | BAC40405.1 | |||
AK143416.1 | BAE25369.1 | |||
AK171667.1 | BAE42599.1 | |||
AK185680.1 | ||||
AK195016.1 | ||||
AK205753.1 | ||||
AK209084.1 | ||||
AK210112.1 | ||||
AK210744.1 | ||||
AK219868.1 | ||||
AK219938.1 | ||||
BC058630.1 | AAH58630.1 | |||
BC138200.1 | ||||
BC145984.1 | ||||
D10576.1 | BAA01433.1 |
More Information
Set Membership: |
Intragenic Non-exonic |