Assay Details
Target Gene Details
Entrez Gene ID: | 22290 |
Gene Name: | ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome |
Gene Aliases: |
Hydb, mKIAA4057 |
Location: |
Chr.Y:1085187-1245754 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uty | NM_009484.2 | 21 | 2947 | NP_033510.2 |
AF057367.1 | 21 | 2927 | AAC67385.1 | |
AK142512.1 | 21 | 2863 | BAE25094.1 | |
AK162410.1 | 19 | 2520 | BAE36901.1 | |
AK164917.1 | 8 | 1392 | BAE37965.1 | |
AK220463.1 | 22 | 2904 | BAD90490.1 |
More Information
Set Membership: |
Intragenic Exonic |