Assay Details
Target Gene Details
Entrez Gene ID: | 23963 |
Gene Name: | teneurin transmembrane protein 1 |
Gene Aliases: |
Odz1, Odz2, TCAP-1, Ten-m1, ten-1 |
Location: |
Chr.X:42527866-43429156 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tenm1 | NM_011855.4 | NP_035985.2 | ||
AB025410.1 | BAA77396.1 | |||
BC138860.1 | ||||
BC138861.1 |