Assay Details
Target Gene Details
Entrez Gene ID: | 20937 |
Gene Name: | suppressor of variegation 3-9 homolog 1 (Drosophila) |
Gene Aliases: |
AI852103, AL022883, DXHXS7466e, H3-K9-HMTase 1, KMT1A, mIS6 |
Location: |
Chr.X:8061171-8074760 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Suv39h1 | NM_001290716.1 | 5 | 1221 | NP_001277645.1 |
NM_011514.2 | 5 | 1240 | NP_035644.1 | |
AF019969.1 | 5 | 1066 | AAB92225.1 | |
AF193861.1 | 5 | 1062 | AAF60969.1 | |
AF193862.1 | 5 | 1165 | AAF60970.1 | |
AK088405.1 | 5 | 1057 | BAC40334.1 | |
AK132238.1 | 4 | 1187 | ||
AK169389.1 | 5 | 1048 | BAE41136.1 | |
AK169592.1 | 5 | 1210 | ||
BC023860.1 | 5 | 1117 | AAH23860.1 |
More Information
Set Membership: |
Intragenic Exonic |