Assay Details
Target Gene Details
Entrez Gene ID: | 73738 |
Gene Name: | HAUS augmin-like complex, subunit 7 |
Gene Aliases: |
1110020L19Rik, Uchl5ip, Uip1 |
Location: |
Chr.X:73437315-73459029 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Haus7 | NM_028633.3 | NP_082909.1 | ||
NM_207104.1 | NP_996987.1 | |||
AK003871.1 | ||||
AK050729.1 | BAC34395.1 | |||
AY029337.1 | AAK38738.1 | |||
BC025464.1 | AAH25464.1 |