Assay Details
Target Gene Details
Entrez Gene ID: | 54646 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3F |
Gene Aliases: |
DXImx48e, R3F, RF3, Sfc15 |
Location: |
Chr.X:7558562-7574281 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r3f | NM_001290574.1 | NP_001277503.1 | ||
NM_138605.3 | NP_613071.3 | |||
AF229644.1 | AAF66954.2 | |||
BC059275.1 | AAH59275.1 | |||
BC138137.1 | ||||
BC138138.1 |
More Information
Set Membership: |
Intragenic Non-exonic |