Assay Details
Target Gene Details
Entrez Gene ID: | 55936 |
Gene Name: | cytidine 5'-triphosphate synthase 2 |
Gene Aliases: |
A830031M15Rik, AI326475, Ctpsh |
Location: |
Chr.X:162901243-163034541 on Build GRCm38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctps2 | NM_001168568.1 | NP_001162040.1 | ||
NM_001168569.1 | NP_001162041.1 | |||
NM_001168571.1 | NP_001162043.1 | |||
NM_018737.5 | NP_061207.1 | |||
NR_033143.1 | ||||
AK032018.1 | BAC27651.1 | |||
AK052854.1 | BAC35174.1 | |||
AK084595.1 | BAC39223.1 | |||
AK085381.1 | ||||
AK150966.1 | BAE29997.1 | |||
AK151597.1 | BAE30537.1 | |||
AK161564.1 | BAE36466.1 | |||
AK164206.1 | BAE37683.1 | |||
BC003257.1 | AAH03257.1 | |||
U49385.1 | AAB17729.1 |
Target Gene Details
Entrez Gene ID: | 12309 |
Gene Name: | S100 calcium binding protein G |
Gene Aliases: |
AU042539, CABP1, CaBP-D9K, Cabp9k, Calb3 |
Location: |
Chr.X:162961992-162964599 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100g | NM_009789.2 | 3 | 359 | NP_033919.1 |
AF028071.1 | 3 | 325 | AAB84041.1 | |
AF136283.1 | 3 | 322 | AAF74818.1 | |
AF293948.1 | 3 | 359 | AAM74560.1 | |
AF293949.1 | 3 | 362 | AAM74561.1 | |
AK146558.1 | 3 | 333 | BAE27259.1 | |
BC010751.1 | 3 | 311 | AAH10751.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |