Assay Details
Target Gene Details
Entrez Gene ID: | 20371 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
JM2, scurfin, sf |
Location: |
Chr.X:7579676-7595243 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxp3 | NM_001199347.1 | NP_001186276.1 | ||
NM_001199348.1 | NP_001186277.1 | |||
NM_054039.2 | NP_473380.1 | |||
AF277991.1 | AAG53605.1 | |||
AF277992.1 | AAG53606.1 | |||
AK157447.1 | BAE34089.1 | |||
AY357712.1 | AAR11305.1 | |||
AY357713.1 | AAR11306.1 | |||
BC132333.1 | ||||
BC132335.1 | ||||
DQ387959.1 | ||||
KF471319.1 |