Assay Details
Target Gene Details
Entrez Gene ID: | 12728 |
Gene Name: | chloride channel 5 |
Gene Aliases: |
5430408K11Rik, ClC-5, Clc5, D930009B12Rik, DXImx42e, Sfc13, T25545 |
Location: |
Chr.X:7153810-7319359 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clcn5 | NM_001243762.1 | NP_001230691.1 | ||
NM_016691.4 | NP_057900.3 | |||
AF134117.1 | AAD28473.1 | |||
AK052990.1 | BAC35233.1 | |||
AK142227.1 | ||||
AK145763.1 | BAE26635.1 | |||
BC036347.1 | AAH36347.1 |
More Information
Set Membership: |
Intragenic Non-exonic |