Assay Details
Target Gene Details
Entrez Gene ID: | 12728 |
Gene Name: | chloride channel 5 |
Gene Aliases: |
5430408K11Rik, ClC-5, Clc5, D930009B12Rik, DXImx42e, Sfc13, T25545 |
Location: |
Chr.X:7153810-7319359 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clcn5 | NM_001243762.1 | 13 | 2653 | NP_001230691.1 |
NM_016691.4 | 11 | 2271 | NP_057900.3 | |
AF134117.1 | 11 | 2271 | AAD28473.1 | |
AK142227.1 | 11 | 2203 | ||
AK145763.1 | 11 | 2240 | BAE26635.1 | |
BC036347.1 | 11 | 2243 | AAH36347.1 |
More Information
Set Membership: |
Intragenic Exonic |