Assay Details
Target Gene Details
Entrez Gene ID: | 26905 |
Gene Name: | eukaryotic translation initiation factor 2, subunit 3, structural gene X-linked |
Gene Aliases: |
AA409828, AA547477, AI314668, Eif-2gx |
Location: |
Chr.X:94188709-94212651 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif2s3x | NM_012010.3 | NP_036140.1 | ||
AJ006587.1 | CAA07099.1 | |||
AK010819.1 | BAB27202.1 | |||
AK128949.1 | ||||
AK146900.1 | BAE27514.1 | |||
AK150095.1 | BAE29303.1 | |||
AK151579.1 | BAE30519.1 | |||
AK153568.1 | BAE32102.1 | |||
AK165866.1 | BAE38425.1 | |||
AK171317.1 | ||||
AK208120.1 | ||||
BC063755.1 | AAH63755.1 | |||
BC139234.1 | ||||
BC139235.1 |
More Information
Set Membership: |
Intragenic Non-exonic |