Assay Details
Target Gene Details
Entrez Gene ID: | 66235 |
Gene Name: | eukaryotic translation initiation factor 1A, X-linked |
Gene Aliases: |
1500010B24Rik, AI426898, Eif1ay |
Location: |
Chr.X:159372195-159385699 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif1ax | NM_025437.4 | 1 | 147 | NP_079713.2 |
AK005185.1 | 1 | 135 | BAB23869.1 | |
AK007301.1 | 1 | 98 | BAB24942.1 | |
AK012233.1 | 1 | 127 | BAB28110.1 | |
AK012464.1 | 1 | 48 | BAB28259.3 | |
AK012714.1 | 1 | 130 | BAB28428.1 | |
AK021283.1 | 1 | 128 | BAB32361.1 | |
AK077710.1 | 1 | 96 | BAC36971.1 | |
AK166629.1 | 1 | 150 | BAE38904.1 | |
AK166852.1 | 1 | 148 | BAE39071.1 | |
BC027284.1 | 1 | 84 | AAH27284.1 |
More Information
Set Membership: |
Intragenic Exonic |