Assay Details
Target Gene Details
Entrez Gene ID: | 320376 |
Gene Name: | BCL6 co-repressor-like 1 |
Gene Aliases: |
6720425J07Rik, B930011H20Rik |
Location: |
Chr.X:48341358-48408571 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcorl1 | NM_178782.4 | NP_848897.3 | ||
AK020114.1 | ||||
CJ176997.1 |