Assay Details
Target Gene Details
Entrez Gene ID: | 27061 |
Gene Name: | B cell receptor associated protein 31 |
Gene Aliases: |
Bap31 |
Location: |
Chr.X:73686178-73716466 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcap31 | NM_012060.4 | NP_036190.2 | ||
AK008043.1 | BAB25427.1 | |||
AK011500.1 | BAB27660.1 | |||
AK151641.1 | BAE30571.1 | |||
BC002106.1 | AAH02106.1 | |||
X81816.1 | CAA57414.1 |
More Information
Set Membership: |
Intragenic Non-exonic |