Assay Details
Target Gene Details
Entrez Gene ID: | 17995 |
Gene Name: | NADH dehydrogenase (ubiquinone) flavoprotein 1 |
Gene Aliases: |
CI-51kD |
Location: |
Chr.19:4007499-4012755 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufv1 | NM_133666.3 | 6 | 986 | NP_598427.1 |
AK005021.1 | 6 | 936 | ||
AK075692.1 | 6 | 931 | BAC35893.1 | |
AK151455.1 | 6 | 897 | ||
AK178320.1 | 2 | 174 | ||
AK185238.1 | 2 | 174 | ||
AK187850.1 | 2 | 174 | ||
AK188080.1 | 1 | 69 | ||
AK188929.1 | 1 | 56 | ||
AK189786.1 | 1 | 174 | ||
AK205680.1 | ||||
AK209435.1 | 2 | 174 | ||
AK210493.1 | 2 | 174 | ||
AK218408.1 | 2 | 174 | ||
BC014818.1 | 6 | 956 | AAH14818.1 | |
BC041682.1 | 6 | 889 | AAH41682.1 |
More Information
Set Membership: |
Intragenic Exonic |