Assay Details
Target Gene Details
Entrez Gene ID: | 226026 |
Gene Name: | structural maintenance of chromosomes 5 |
Gene Aliases: |
MLZ-453, SMC-5, Smc5l1, mKIAA0594, mSMC5 |
Location: |
Chr.19:23206441-23273908 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc5 | NM_001252684.1 | 4 | 549 | NP_001239613.1 |
NM_001252685.1 | 4 | 378 | NP_001239614.1 | |
NM_153808.2 | 4 | 549 | NP_722503.1 | |
AJ534941.1 | 4 | 470 | CAD59184.1 | |
AK143818.1 | 4 | 526 | ||
AK161254.1 | 4 | 379 | ||
BC038345.1 | 4 | 529 | AAH38345.1 | |
CN530965.1 | 1 | 71 |