Assay Details
Target Gene Details
Entrez Gene ID: | 226026 |
Gene Name: | structural maintenance of chromosomes 5 |
Gene Aliases: |
MLZ-453, SMC-5, Smc5l1, mKIAA0594, mSMC5 |
Location: |
Chr.19:23206441-23273908 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc5 | NM_001252684.1 | 25 | 3402 | NP_001239613.1 |
NM_001252685.1 | 24 | 3189 | NP_001239614.1 | |
NM_153808.2 | 24 | 3360 | NP_722503.1 | |
AJ534941.1 | CAD59184.1 | |||
AK049407.1 | 2 | 274 | ||
AK122321.1 | 17 | 5553 | BAC65603.3 | |
AK161254.1 | 24 | 3192 | ||
BC007174.1 | 16 | 1958 | AAH07174.1 | |
BC038345.1 | 24 | 3340 | AAH38345.1 |