Assay Details
Target Gene Details
Entrez Gene ID: | 381218 |
Gene Name: | RIKEN cDNA 4430402I18 gene |
Gene Aliases: |
AW122618, Spata6l |
Location: |
Chr.19:28845528-28970888 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4430402I18Rik |
Target Gene Details
Entrez Gene ID: | 20510 |
Gene Name: | solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1 |
Gene Aliases: |
D130048G10Rik, EAAC1, EAAC2, EAAT3, MEAAC1 |
Location: |
Chr.19:28835166-28913960 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc1a1 | NM_009199.2 | 9 | 1009 | NP_033225.1 |
AK018188.1 | 9 | 1042 | BAB31114.1 | |
AK045632.1 | 9 | 1041 | ||
AK083097.1 | 9 | 1044 | ||
BC064797.1 | 9 | 1031 | AAH64797.1 | |
BC065099.1 | 9 | 1033 | AAH65099.1 | |
D43797.1 | 9 | 1015 | BAA07855.1 | |
U73521.1 | 9 | 1009 | AAB80694.1 | |
U75214.1 | 5 | 784 | AAB71739.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |