Assay Details
Target Gene Details
Entrez Gene ID: | 67264 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8 |
Gene Aliases: |
2900010I05Rik, AI987932 |
Location: |
Chr.19:44548576-44555446 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufb8 | NM_026061.2 | NP_080337.1 | ||
AK010177.1 | BAB26749.1 | |||
AK013516.1 | BAB28893.1 | |||
AK028070.1 | BAE43264.1 | |||
AK206286.1 | ||||
BC043019.1 | AAH43019.1 |
More Information
Set Membership: |
Intragenic Non-exonic |