Assay Details
Target Gene Details
Entrez Gene ID: | 65221 |
Gene Name: | solute carrier family 15, member 3 |
Gene Aliases: |
Ci1, cI-1 |
Location: |
Chr.19:10842544-10857915 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc15a3 | NM_023044.2 | NP_075531.2 | ||
AF121080.1 | AAD24570.1 | |||
AK051941.1 | BAC34816.1 | |||
BC051940.1 | AAH51940.1 |
More Information
Set Membership: |
Intragenic Non-exonic |