Assay Details
Target Gene Details
Entrez Gene ID: | 69060 |
Gene Name: | pancreatic lipase |
Gene Aliases: |
1810007A24Rik, PL, PTL |
Location: |
Chr.19:58670365-58681788 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnlip | NM_026925.3 | 12 | 1242 | NP_081201.2 |
AY387690.1 | 11 | 1214 | AAQ90020.1 | |
BC061061.1 | 12 | 1242 | AAH61061.1 |
More Information
Set Membership: |
Intragenic Exonic |