Assay Details
Target Gene Details
Entrez Gene ID: | 18563 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
Pc, Pcb |
Location: |
Chr.19:4510472-4621752 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcx | NM_001162946.1 | 15 | 2283 | NP_001156418.1 |
NM_008797.3 | 14 | 2260 | NP_032823.2 | |
AK077665.1 | 6 | 1088 | BAC36939.1 | |
AK148328.1 | 15 | 2230 | BAE28484.1 | |
AK170596.1 | 15 | 2243 | BAE41902.1 | |
AK170871.1 | 15 | 2233 | ||
AK172320.1 | 15 | 2211 | BAE42943.1 | |
AK201626.1 | ||||
AK206273.1 | ||||
AK210822.1 | ||||
AK212052.1 | ||||
AK213300.1 | ||||
AK217110.1 | ||||
BC055030.1 | 15 | 2218 | AAH55030.1 | |
L09192.1 | 14 | 2236 | AAA39737.1 | |
M97957.1 | 9 | 1408 | AAA39897.1 |
More Information
Set Membership: |
Intragenic Exonic |