Assay Details
Target Gene Details
Entrez Gene ID: | 54683 |
Gene Name: | peroxiredoxin 5 |
Gene Aliases: |
AOEB166, AOPP, Pmp20, Prdx6, PrxV |
Location: |
Chr.19:6906819-6909645 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prdx5 | NM_012021.2 | NP_036151.1 | ||
AB041573.1 | BAA95057.1 | |||
AF110733.1 | AAG13450.1 | |||
AF124994.1 | AAF27532.1 | |||
AF197951.1 | AAF04855.1 | |||
AK002383.1 | BAB22058.1 | |||
AK003332.2 | BAB22720.1 | |||
AK009858.1 | BAB26548.1 | |||
AK136130.1 | BAE22835.1 | |||
AK152653.1 | BAE31390.1 | |||
BC008174.1 | AAH08174.1 | |||
BC014682.1 | AAH14682.1 |
More Information
Set Membership: |
Intragenic Non-exonic |