Assay Details
Target Gene Details
Entrez Gene ID: | 107765 |
Gene Name: | ankyrin repeat domain 1 (cardiac muscle) |
Gene Aliases: |
Alrp, CARP, Crap, MARP1 |
Location: |
Chr.19:36111965-36119844 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ankrd1 | NM_013468.3 | 2 | 220 | NP_038496.2 |
AF041847.1 | 2 | 171 | AAC03533.1 | |
AK009655.1 | 2 | 213 | BAB26419.1 | |
AK009959.1 | 2 | 219 | BAB26611.1 | |
AK145944.1 | 2 | 261 | BAE26773.1 | |
AK146471.1 | 2 | 217 | BAE27197.1 | |
AK146627.1 | 2 | 219 | BAE27316.1 | |
AK146940.1 | 2 | 217 | BAE27549.1 | |
BC037138.1 | 2 | 220 | AAH37138.1 |