Assay Details
Target Gene Details
Entrez Gene ID: | 64697 |
Gene Name: | kidney expressed gene 1 |
Gene Aliases: |
0610008P16Rik, AI316519, GS4059 |
Location: |
Chr.19:12695790-12719902 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Keg1 | NM_029550.4 | NP_083826.1 | ||
AB028071.1 | ||||
AK002360.1 | BAB22039.1 | |||
AK165349.1 | ||||
AK165462.1 | 3 | 1677 | ||
AK165515.1 | BAE38231.1 | |||
BC010803.1 | AAH10803.1 |
More Information
Set Membership: |
Intragenic Non-exonic |