Assay Details
Target Gene Details
Entrez Gene ID: | 11632 |
Gene Name: | aryl-hydrocarbon receptor-interacting protein |
Gene Aliases: |
AA408703, AW476050, Ara9, D19Bwg1412e, Fkbp16, Xap2 |
Location: |
Chr.19:4113756-4127236 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aip | NM_001276284.1 | 3 | 555 | NP_001263213.1 |
NM_016666.3 | 4 | 654 | NP_057875.1 | |
AK011609.1 | 3 | 428 | ||
AK146390.1 | 3 | 557 | BAE27133.1 | |
BC075614.1 | 3 | 426 | AAH75614.1 | |
CX222057.1 | 3 | 524 | ||
U85489.1 | 4 | 623 | AAB59009.1 |
More Information
Set Membership: |
Intragenic Exonic |