Assay Details
Target Gene Details
Entrez Gene ID: | 226016 |
Gene Name: | abhydrolase domain containing 17B |
Gene Aliases: |
5730446C15Rik, Cgi67, Fam108b, Fam108b1 |
Location: |
Chr.19:21653309-21685638 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abhd17b | NM_146096.3 | NP_666208.2 | ||
AK050879.1 | ||||
AK152807.1 | 1 | 64 | BAE31512.1 | |
BC032261.1 | 1 | 293 | AAH32261.1 | |
BC091733.1 | AAH91733.1 |
More Information
Set Membership: |
Intragenic Non-exonic |