Assay Details
Target Gene Details
Entrez Gene ID: | 14537 |
Gene Name: | glucosaminyl (N-acetyl) transferase 1, core 2 |
Gene Aliases: |
5630400D21Rik, B130048E03, C2 GlcNAcT, C2GNT, IGnT |
Location: |
Chr.19:17326141-17372844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gcnt1 | NM_001136484.3 | NP_001129956.3 | ||
NM_010265.5 | NP_034395.4 | |||
NM_173442.5 | NP_775618.3 | |||
AK045216.1 | BAC32265.1 | |||
AK167048.1 | ||||
AK199888.1 | ||||
BC039126.1 | AAH39126.1 | |||
BY010642.1 | ||||
BY768580.1 | ||||
CJ092264.1 | ||||
D87332.1 | BAA22998.1 | |||
D87333.1 | BAA22999.1 | |||
U19265.1 | 1 | 56 | AAA60948.1 |
More Information
Set Membership: |
Intragenic Non-exonic |