Assay Details
Target Gene Details
Entrez Gene ID: | 15488 |
Gene Name: | hydroxysteroid (17-beta) dehydrogenase 4 |
Gene Aliases: |
17-beta-HSD, 17[b]-HSD, 4, DBP, MFE-2, MFP2, MPF-2, Mfp-2, perMFE-2 |
Location: |
Chr.18:50128201-50196270 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsd17b4 | NM_008292.4 | NP_032318.2 | ||
AK004866.1 | BAB23627.1 | |||
AK088381.1 | BAC40317.1 | |||
AK147182.1 | BAE27744.1 | |||
AK152532.1 | ||||
AK161655.1 | BAE36514.1 | |||
AK166801.1 | BAE39029.1 | |||
AK167060.1 | BAE39222.1 | |||
AK169077.1 | BAE40862.1 | |||
BC022175.1 | AAH22175.1 | |||
X89998.1 | CAA62015.1 |