Assay Details
Target Gene Details
Entrez Gene ID: | 545253 |
Gene Name: | predicted gene 5820 |
Gene Aliases: |
EG545253 |
Location: |
Chr.18:38892456-38971274 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm5820 | NM_001033789.2 | NP_001028961.1 | ||
AK145364.1 | BAE26390.1 | |||
BC147564.1 |