Assay Details
Target Gene Details
Entrez Gene ID: | 268996 |
Gene Name: | synovial sarcoma translocation, Chromosome 18 |
Gene Aliases: |
D130059H17, Ssxt, Syt |
Location: |
Chr.18:14625629-14683002 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ss18 | NM_001161369.1 | NP_001154841.1 | ||
NM_001161370.1 | NP_001154842.1 | |||
NM_001161371.1 | NP_001154843.1 | |||
NM_009280.2 | NP_033306.2 | |||
AK039430.1 | ||||
AK052476.1 | BAC35009.1 | |||
AK083622.1 | BAC38970.1 | |||
AK083887.1 | BAC39049.1 | |||
AK149325.1 | BAE28814.1 | |||
AK166053.1 | BAE38546.1 | |||
AK179801.1 | ||||
AK183049.1 | ||||
BC065062.1 | AAH65062.1 | |||
BC096742.1 | AAH96742.1 | |||
X93357.1 | CAA63733.1 |
More Information
Set Membership: |
Intragenic Non-exonic |