Assay Details
Target Gene Details
Entrez Gene ID: | 13512 |
Gene Name: | desmoglein 3 |
Gene Aliases: |
bal, balding |
Location: |
Chr.18:20510304-20541310 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dsg3 | NM_030596.3 | NP_085099.2 | ||
AK037142.1 | BAC29718.1 | |||
AK148657.1 | BAE28635.1 | |||
BC132058.1 | ||||
BC132639.1 | ||||
U86016.1 | AAB65091.1 |
More Information
Set Membership: |
Intragenic Non-exonic |