Assay Details
Target Gene Details
Entrez Gene ID: | 26384 |
Gene Name: | glucosamine-6-phosphate deaminase 1 |
Gene Aliases: |
GNPDA, Gnp1, Gnpi, oscillin |
Location: |
Chr.18:38327537-38338993 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnpda1 | NM_011937.2 | NP_036067.2 | ||
AF088903.1 | AAC36739.1 | |||
AF160355.1 | AAD42233.1 | |||
AK080610.1 | ||||
AK084247.1 | ||||
AK166069.1 | ||||
BC014800.1 | AAH14800.1 | |||
BY092839.1 | ||||
CJ048539.1 |
More Information
Set Membership: |
Intragenic Non-exonic |